2-Chloropyridine-3,4-dicarboxylic acid 98%

Supplier: Ambeed
Warning

A179659-1G A179659-100MG A179659-250MG
103807-454EA 308 USD
103807-454 103845-806 77083-276 77260-176 77260-178
2-Chloropyridine-3,4-dicarboxylic acid 98%
2-Chloropyridine-3,4-dicarboxylic acid

Precaución: for Research Use Only

Formula: C₇H₄ClNO₄
Boiling Pt: 524.3 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD01318441
CAS Number: 215306-02-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR