2-Chloropyridine-3,4-diamine 98%

Supplier: Ambeed
Warning

Synonyms: 3,4-Diamino-2-chloropyridine, 2-Chloropyridine-3,4-diamine

A146572-10G A146572-250MG A146572-25G A146572-5G A146572-1G
103803-682EA 7 USD
103803-682 103841-774 76972-868 76972-870 77299-532 77299-534 77317-106
2-Chloropyridine-3,4-diamine 98%
2-Chloropyridine-3,4-diamine

Precaución: for Research Use Only

Formula: C₅H₆ClN₃
MW: 143.5742 g/mol
Boiling Pt: 381.3 °C (760 mm Hg)
Melting Pt: 158…163 °C
Flash Pt: 184.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07363739
CAS Number: 39217-08-8
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR