3-Amino-2-chloropyridine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-3-pyridylamine, 2-Chloropyridin-3-amine

A251091-100G A251091-10G A251091-500G A251091-25G A251091-5G A251091-1000G
103809-784EA 215 USD
103809-784 103851-802 77013-996 77013-998 77014-000 77091-170 77091-172 77304-548
3-Amino-2-chloropyridine 98%
3-Amino-2-chloropyridine

Precaución: for Research Use Only

Formula: C₅H₅ClN₂
MW: 128.56 g/mol
Boiling Pt: 262.8 °C (760 mm Hg)
Melting Pt: 76…78 °C
Flash Pt: 185 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006238
CAS Number: 6298-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR