2-Chloro-N-phenylacetamide 95%

Supplier: Ambeed
Warning

A119644-250MG A119644-25G A119644-1G A119644-10G
103799-810EA 13 USD
103799-810 77258-048 77258-050 77311-918 77747-870
2-Chloro-N-phenylacetamide 95%
2-Chloro-N-phenylacetamide

Precaución: for Research Use Only

Formula: C₈H₈ClNO
Melting Pt: 136…139 °C
Flash Pt: 159.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00018887
CAS Number: 587-65-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR