2-Chloro-N-methylpyrimidin-4-amine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Chloro-4-(methylamino)pyrimidine
A125024-1G
A125024-5G
A125024-250MG
103800-606EA
22
USD
103800-606
77312-754
77312-756
77624-044
2-Chloro-N-methylpyrimidin-4-amine 97%
2-Chloro-N-methylpyrimidin-4-amine
Precaución: for Research Use Only
Formula:
C₅H₆ClN₃ MW: 143.58 g/mol Boiling Pt: 317.228 °C (760 mm Hg) Flash Pt: 145.655 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09055368 CAS Number: 66131-68-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...