2-Chloro-N-methoxy-N-methylacetamide 98%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: Methyl 2-Chloro-N-methylacetohydroxamate

A100718-10G A100718-25G A100718-5G A100718-100G A100718-500G
103821-290EA 5 USD
103821-290 76821-212 76821-214 77282-550 77548-898 77682-933
2-Chloro-N-methoxy-N-methylacetamide 98%, Ambeed.Inc
2-Chloro-N-methoxy-N-methylacetamide

Precaución: For research use only.

Formula: C₄H₈ClNO₂
MW: 137.56 g/mol
Boiling Pt: 141.5 °C (760 mm Hg)
Melting Pt: 39…41 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00134232
CAS Number: 67442-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR