2-Chloro-N-4-methylpyridine-3,4-diamine 95%

Supplier: Ambeed
Warning

A154302-100MG A154302-250MG A154302-1G A154302-5G
103804-622EA 99 USD
103804-622 103842-808 77259-390 77259-392 77300-048 77300-050
2-Chloro-N-4-methylpyridine-3,4-diamine 95%
2-Chloro-N-4-methylpyridine-3,4-diamine

Precaución: for Research Use Only

Formula: C₆H₈ClN₃
Storage Temperature: Refrigerator
MDL Number: MFCD00778574
CAS Number: 50432-67-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR