2-Chloro-N-(2,2,2-trifluoroethyl)acetamide 98%

Supplier: Ambeed
Danger

A130466-100MG A130466-10G A130466-250MG A130466-1G
103801-426EA 59 USD
103801-426 76823-002 76823-004 76823-006 77313-860
2-Chloro-N-(2,2,2-trifluoroethyl)acetamide 98%
2-Chloro-N-(2,2,2-trifluoroethyl)acetamide

Precaución: for Research Use Only

Formula: C₄H₅ClF₃NO
MW: 175.537 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03984449
CAS Number: 170655-44-4
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR