2-Chloroisonicotinic acid 98% (contains < 5% Salt)

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-4-pyridinecarboxylic acid, 2-Chloropyridine-4-carboxylic acid

A109437-100G A109437-10G A109437-25G A109437-500G A109437-5G
103823-302EA 5 USD
103823-302 77093-892 77093-894 77093-896 77093-898 77310-118
2-Chloroisonicotinic acid 98% (contains < 5% Salt)
2-Chloroisonicotinic acid

Precaución: for Research Use Only

Formula: C₆H₄ClNO₂
MW: 157.56 g/mol
Boiling Pt: 417.7 °C (760 mm Hg)
Melting Pt: 246 °C
Flash Pt: 206.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00191402
CAS Number: 6313-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR