2-Chloro-4-formylpyridine 97%

Supplier: Ambeed
Warning

Synonyms: 2-Chloropyridine-4-carboxaldehyde, 2-Chloroisonicotinaldehyde

A532317-25G A532317-5G A532317-100G A532317-10G A532317-250MG A532317-1G A532317-500G
103828-576EA 8 USD
103828-576 103839-688 76843-704 76843-706 77103-320 77103-322 77103-324 77307-748 77686-375
2-Chloro-4-formylpyridine 97%
2-Chloro-4-formylpyridine

Precaución: for Research Use Only

Formula: C₆H₄ClNO
MW: 141.56 g/mol
Melting Pt: 46…50 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06651557
CAS Number: 101066-61-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR