2-Chlorodibenzo[b,f][1,4]thiazepin-11(10H)-one 97%

Supplier: Ambeed
Warning

A196447-100MG A196447-250MG A196447-1G
103816-846EA 52 USD
103816-846 76814-110 76814-112 77085-414
2-Chlorodibenzo[b,f][1,4]thiazepin-11(10H)-one 97%
2-Chlorodibenzo[b,f][1,4]thiazepin-11(10H)-one

Precaución: for Research Use Only

Formula: C₁₃H₈ClNOS
MW: 261.73 g/mol
Boiling Pt: 351.3±41.0 °C (760 mm Hg)
Melting Pt: 250…252 °C
Flash Pt: 351.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD22495113
CAS Number: 3159-04-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR