2-Chlorocinnamic acid 99%

Supplier: Ambeed
Warning

A299449-100G A299449-10G A299449-500G A299449-25G A299449-5G
103835-376EA 5 USD
103835-376 76693-382 76693-384 76693-386 77305-766 77305-768
2-Chlorocinnamic acid 99%
2-Chlorocinnamic acid

Precaución: for Research Use Only

Formula: C₉H₇ClO₂
Boiling Pt: 321.6 °C (760 mm Hg)
Melting Pt: 209…212 °C (760 mm Hg)
Flash Pt: 148.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00004372
CAS Number: 3752-25-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR