2-Chloroadenosine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(2R,3R,4S,5R)-2-(6-Amino-2-chloro-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol
A209676-1G
A209676-25G
A209676-5G
A209676-250MG
A209676-100G
103817-952EA
6
USD
103817-952
103848-638
77086-988
77086-990
77086-992
77275-128
77748-311
2-Chloroadenosine 98%
2-Chloroadenosine
Precaución: for Research Use Only
Formula:
C₁₀H₁₂ClN₅O₄ MW: 301.69 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00005734 CAS Number: 146-77-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...