2-Chloro-9H-purine 97%

Supplier: Ambeed
Warning

A114127-100MG A114127-250MG
103798-352EA 42 USD
103798-352 77257-820 77257-822
2-Chloro-9H-purine 97%
2-Chloro-9H-purine

Precaución: for Research Use Only

Formula: C₅H₃ClN₄
MW: 154.5600 g/mol
Boiling Pt: 397.7 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00137826
CAS Number: 1681-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR