2-Chloro-8-methoxyquinoxaline 95%

Supplier: Ambeed
Warning

A184796-100MG A184796-250MG
103815-784EA 150 USD
103815-784 77164-016 77164-018
2-Chloro-8-methoxyquinoxaline 95%
2-Chloro-8-methoxyquinoxaline

Precaución: Inert atmosphere, 2…8 °C.

Formula: C₉H₇ClN₂O
MW: 194.618 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD25368345
CAS Number: 659729-70-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR