2-Chloro-7-methyl-1H-benzo[d]imidazole 95%

Supplier: Ambeed
Warning

A133953-100MG A133953-250MG
103801-940EA 147 USD
103801-940 77234-630 77234-632
2-Chloro-7-methyl-1H-benzo[d]imidazole 95%
2-Chloro-7-methyl-1H-benzo[d]imidazole

Precaución: for Research Use Only

Formula: C₈H₇ClN₂
MW: 166.61 g/mol
Boiling Pt: 339.9±35.0 °C (760 mm Hg)
Flash Pt: 189.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10001473
CAS Number: 15965-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR