2-Chloro-7H-purin-6-amine 97%

Supplier: Ambeed
Warning

Synonyms: 6-Amino-2-chloropurine, 2-Chloroadenine

A122294-100G A122294-10G A122294-25G A122294-1G A122294-5G
103800-200EA 6 USD
103800-200 77284-864 77284-866 77284-868 77312-334 77312-336
2-Chloro-7H-purin-6-amine 97%
2-Chloro-7H-purin-6-amine

Precaución: for Research Use Only

Formula: C₅H₄ClN₅
MW: 169.57 g/mol
Boiling Pt: 315.2 °C (760 mm Hg)
Flash Pt: 144.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00037925
CAS Number: 1839-18-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR