2-Chloro-6-phenylpyrimidin-4-amine 95%

Supplier: Ambeed
Warning

A125083-250MG A125083-1G
103800-624EA 364 USD
103800-624 76983-430 77312-772
2-Chloro-6-phenylpyrimidin-4-amine 95%
2-Chloro-6-phenylpyrimidin-4-amine

Precaución: for Research Use Only

Formula: C₁₀H₈ClN₃
MW: 205.64 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16249541
CAS Number: 54994-35-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR