2-Chloro-6-nitrobenzoic acid 98%

Supplier: Ambeed
Warning

A137181-100G A137181-5G A137181-25G
77137-166EA 49.87 USD
77137-166 77298-898 77315-378
2-Chloro-6-nitrobenzoic acid 98%
2-Chloro-6-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄ClNO₄
MW: 201.57 g/mol
Boiling Pt: 345.7 °C (760 mm Hg)
Melting Pt: 164…166 °C
Storage Temperature: Ambient
MDL Number: MFCD00100425
CAS Number: 5344-49-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR