2-Chloro-6-methylpyridine 98%

Supplier: Ambeed
Warning

Synonyms: 6-Chloro-2-picoline

A124905-100G A124905-10G A124905-25G
77285-066EA 31 USD
77285-066 77285-068 77285-070 77312-740
2-Chloro-6-methylpyridine 98%
2-Chloro-6-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₆ClN
MW: 127.57 g/mol
Boiling Pt: 64
Flash Pt: 74 °C
Storage Temperature: Ambient
MDL Number: MFCD00006245
CAS Number: 18368-63-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR