2-Chloro-6-methylpyridin-4-amine 97%
Supplier: Ambeed
|
Warning
|
A184221-10G
A184221-250MG
A184221-1G
A184221-25G
103815-728EA
25
USD
103815-728
103846-290
77029-572
77029-574
77083-902
77083-904
2-Chloro-6-methylpyridin-4-amine 97%
2-Chloro-6-methylpyridin-4-amine
Precaución: for Research Use Only
Formula:
C₆H₇ClN₂ MW: 142.586 g/mol Melting Pt: 155…157 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00235132 CAS Number: 79055-63-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...