2-Chloro-6-methylpyrazine 98%

Supplier: Ambeed
Warning

A268512-10G A268512-1G
103812-916EA 9 USD
103812-916 103852-818 77262-406 77305-004
2-Chloro-6-methylpyrazine 98%
2-Chloro-6-methylpyrazine

Precaución: for Research Use Only

Formula: C₅H₅ClN₂
MW: 128.5596 g/mol
Boiling Pt: 173.6 °C (760 mm Hg)
Melting Pt: 49…52 °C
Flash Pt: 73.337 °C
Storage Temperature: Ambient
MDL Number: MFCD00055032
CAS Number: 38557-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR