2-Chloro-6-methylnicotinic acid 98%

Supplier: Ambeed
Warning

A181527-5G A181527-100G A181527-10G A181527-25G A181527-1G
103807-614EA 9 USD
103807-614 103845-988 77083-512 77138-738 77138-740 77138-742 77301-722
2-Chloro-6-methylnicotinic acid 98%
2-Chloro-6-methylnicotinic acid

Precaución: for Research Use Only

Formula: C₇H₆ClNO₂
Boiling Pt: 325.5 °C (760 mm Hg)
Melting Pt: 162…164 °C
Flash Pt: 150.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00134165
CAS Number: 30529-70-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR