2-Chloro-6-methylisonicotinic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-6-methylpyridine-4-carboxylic acid

A108650-100G A108650-10G A108650-250MG A108650-25G A108650-500G A108650-5G A108650-1G
103797-378EA 9 USD
103797-378 103823-160 76995-062 76995-064 76995-066 76995-068 76995-070 77283-766 77309-998
2-Chloro-6-methylisonicotinic acid 98%
2-Chloro-6-methylisonicotinic acid

Precaución: for Research Use Only

Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Boiling Pt: 399 °C (760 mm Hg)
Melting Pt: 208…212 °C
Flash Pt: 195.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00052830
CAS Number: 25462-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR