6-Chloro-o-toluidine 98%

Supplier: Ambeed
Warning

A273583-100G A273583-10G A273583-500G A273583-5G A273583-25G
103853-068EA 6 USD
103853-068 77031-980 77031-982 77031-984 77092-864 77305-140
6-Chloro-o-toluidine 98%
6-Chloro-o-toluidine

Precaución: for Research Use Only

Formula: C₇H₈ClN
Boiling Pt: 215 °C (760 mm Hg)
Melting Pt: 10…12 °C
Flash Pt: 105 °C
Storage Temperature: Ambient
MDL Number: MFCD00007679
CAS Number: 87-63-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR