2-Chloro-6-methoxyphenylboronic acid 97%

Supplier: Ambeed
Warning

A306473-1G A306473-25G A306473-5G A306473-100MG A306473-10G A306473-250MG
103814-470EA 8 USD
103814-470 103835-636 76838-348 76838-350 76838-352 77032-532 77032-534 77032-536
2-Chloro-6-methoxyphenylboronic acid 97%
2-Chloro-6-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BClO₃
MW: 186.4000 g/mol
Boiling Pt: 355.5 °C (760 mm Hg)
Flash Pt: 168.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04039878
CAS Number: 385370-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR