2-Chloro-6-methoxybenzaldehyde 98%

Supplier: Ambeed
Warning

A112595-10G A112595-250MG A112595-25G A112595-1G A112595-5G
103798-050EA 8 USD
103798-050 103823-888 76982-808 76982-810 76982-812 77310-700 77310-702
2-Chloro-6-methoxybenzaldehyde 98%
2-Chloro-6-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇ClO₂
MW: 170.5900 g/mol
Boiling Pt: 263 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD08059233
CAS Number: 29866-54-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR