2-Chloro-6-methoxyaniline 98%
Supplier: Ambeed
|
Warning
|
A533619-100MG
A533619-10G
A533619-250MG
A533619-1G
A533619-25G
A533619-5G
103828-586EA
20
USD
103828-586
103839-698
76878-350
76878-352
76878-354
77307-752
77307-754
77307-756
2-Chloro-6-methoxyaniline 98%
2-Chloro-6-methoxyaniline
Precaución: for Research Use Only
Formula:
C₇H₈ClNO MW: 157.598 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD14455911 CAS Number: 158966-62-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...