2-Chloro-6-iodotoluene 98%

Supplier: Ambeed
Warning

A173578-5G A173578-100G A173578-10G
103845-132EA 7 USD
103845-132 77082-380 77260-000 77260-002
2-Chloro-6-iodotoluene 98%
2-Chloro-6-iodotoluene

Precaución: for Research Use Only

Formula: C₇H₆ClI
Boiling Pt: 243 °C (760 mm Hg)
Melting Pt: 11 °C
Flash Pt: 100.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00019013
CAS Number: 42048-11-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR