2-Chloro-6-iodopyridine 98%
Supplier: Ambeed
|
Warning
|
A247712-1G
A247712-5G
103811-790EA
24
USD
103811-790
103851-626
77090-962
77090-964
2-Chloro-6-iodopyridine 98%
2-Chloro-6-iodopyridine
Precaución: for Research Use Only
Formula:
C₅H₃ClIN Boiling Pt: 255.5 °C (760 mm Hg) Flash Pt: 108.3 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD07784182 CAS Number: 258506-66-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...