6-Chlorosalicylaldehyde 98%

Supplier: Ambeed
Warning

A181530-1G A181530-25G A181530-5G A181530-100G A181530-10G
103807-616EA 11 USD
103807-616 103845-990 77083-514 77083-516 77083-518 77138-744 77138-746
6-Chlorosalicylaldehyde 98%
6-Chlorosalicylaldehyde

Precaución: for Research Use Only

Formula: C₇H₅ClO₂
Boiling Pt: 223.1 °C (760 mm Hg)
Melting Pt: 51…55 °C
Flash Pt: 107 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01646166
CAS Number: 18362-30-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR