2-Chloro-6-hydrazinopyridine 97%

Supplier: Ambeed
Warning

Synonyms: (6-Chloro-2-pyridyl)hydrazine

A175662-25G A175662-5G A175662-100G
103845-386EA 7 USD
103845-386 77082-730 77082-732 77260-074
2-Chloro-6-hydrazinopyridine 97%
2-Chloro-6-hydrazinopyridine

Precaución: for Research Use Only

Formula: C₅H₆ClN₃
Boiling Pt: 308.6 °C (760 mm Hg)
Melting Pt: 118 °C
Storage Temperature: Freezer
MDL Number: MFCD00214724
CAS Number: 5193-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR