2-Chloro-6-formylbenzonitrile 98%

Supplier: Ambeed
Warning

A257398-100MG A257398-250MG A257398-1G A257398-5G
103812-302EA 62 USD
103812-302 103852-170 76827-084 76827-086 77304-702 77304-704
2-Chloro-6-formylbenzonitrile 98%
2-Chloro-6-formylbenzonitrile

Precaución: for Research Use Only

Formula: C₈H₄ClNO
MW: 165.58 g/mol
Boiling Pt: 323.5 °C (760 mm Hg)
Flash Pt: 149.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10699534
CAS Number: 77532-86-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR