2-Chloro-6-fluoroquinoxaline 95%
Supplier: Ambeed
|
Warning
|
A110633-100MG
A110633-250MG
A110633-1G
103797-696EA
44
USD
103797-696
103823-504
76781-052
76781-054
77310-306
2-Chloro-6-fluoroquinoxaline 95%
2-Chloro-6-fluoroquinoxaline
Precaución: for Research Use Only
Formula:
C₈H₄ClFN₂ MW: 182.5800 g/mol Boiling Pt: 257.1 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD09999192 CAS Number: 55687-33-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...