1-Chloro-3-fluoro-2-nitrobenzene 97%
Supplier: Ambeed
|
Warning
|
A276496-25G
A276496-5G
A276496-100G
A276496-10G
A276496-1G
103813-306EA
9
USD
103813-306
103853-240
77093-098
77093-100
77289-546
77289-548
77305-236
1-Chloro-3-fluoro-2-nitrobenzene 97%
1-Chloro-3-fluoro-2-nitrobenzene
Precaución: for Research Use Only
Formula:
C₆H₃ClFNO₂ MW: 175.55 g/mol Boiling Pt: 245.1 °C (760 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD06658264 CAS Number: 64182-61-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...