2-Chloro-6-fluorobenzoic acid 98%
Supplier: Ambeed
|
Warning
|
A762937-100G
A762937-10G
A762937-500G
A762937-1000G
A762937-25G
103810-556EA
142
USD
103810-556
103840-896
77146-330
77146-332
77146-334
77308-274
77308-276
2-Chloro-6-fluorobenzoic acid 98%
2-Chloro-6-fluorobenzoic acid
Precaución: for Research Use Only
Formula:
C₇H₄ClFO₂ MW: 174.56 g/mol Boiling Pt: 263.7 °C (760 mm Hg) Melting Pt: 155…157 °C Flash Pt: 113.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002417 CAS Number: 434-75-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...