2-Chloro-6-fluoroanisole 97%

Supplier: Ambeed
Warning

A231168-1G A231168-5G
103810-758EA 8 USD
103810-758 103850-506 77089-380 77089-382
2-Chloro-6-fluoroanisole 97%
2-Chloro-6-fluoroanisole

Precaución: for Research Use Only

Formula: C₇H₆ClFO
MW: 160.58 g/mol
Boiling Pt: 186.4 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD01631573
CAS Number: 53145-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR