2-Chloro-6,7-dihydro-5H-cyclopenta[b]pyridine 95%

Supplier: Ambeed
Warning

A139767-100MG A139767-250MG A139767-1G
103802-772EA 57 USD
103802-772 77027-430 77027-432 77299-090
2-Chloro-6,7-dihydro-5H-cyclopenta[b]pyridine 95%
2-Chloro-6,7-dihydro-5H-cyclopenta[b]pyridine

Precaución: for Research Use Only

Formula: C₈H₈ClN
MW: 153.61 g/mol
Boiling Pt: 236.5±40.0 °C (760 mm Hg)
Flash Pt: 119.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09034995
CAS Number: 117890-55-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR