2-Chloro-6-(trifluoromethyl)pyridine 98%

Supplier: Ambeed
Warning

A702485-25G A702485-100G A702485-10G A702485-500G
103840-652EA 6 USD
103840-652 76844-866 77280-838 77280-840 77280-842
2-Chloro-6-(trifluoromethyl)pyridine 98%
2-Chloro-6-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₃ClF₃N
MW: 181.54
Boiling Pt: 147.5 °C (760 mm Hg)
Melting Pt: 33…35 °C
Storage Temperature: Ambient
MDL Number: MFCD00042226
CAS Number: 39890-95-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR