2-Chloro-6-(trifluoromethoxy)pyridine 95%

Supplier: Ambeed
Warning

A191630-100MG A191630-10G A191630-250MG A191630-1G A191630-5G
103816-388EA 98 USD
103816-388 103846-990 76873-090 76873-092 76873-094 77084-836 77084-838
2-Chloro-6-(trifluoromethoxy)pyridine 95%
2-Chloro-6-(trifluoromethoxy)pyridine

Precaución: for Research Use Only

Formula: C₆H₃ClF₃NO
MW: 197.542 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18909427
CAS Number: 1221171-70-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR