5-Chlorothiophene-2-carboxaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-5-thiophenecarboxaldehyde, 2-Chloro-5-formylthiophene

A128562-100G A128562-10G A128562-25G A128562-5G
76983-562EA 153.84 USD
76983-562 76983-564 77313-484 77313-486
5-Chlorothiophene-2-carboxaldehyde 97%
5-Chlorothiophene-2-carboxaldehyde

Precaución: for Research Use Only

Formula: C₅H₃ClOS
MW: 146.59 g/mol
Boiling Pt: 221.1 °C (760 mm Hg)
Flash Pt: 87.5 °C
Storage Temperature: Freezer
MDL Number: MFCD00047090
CAS Number: 7283-96-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR