2-Chloro-5-pyrimidineboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Chloropyrimidine-5-boronic acid

A110557-10G A110557-25G A110557-1G
103797-682EA 9 USD
103797-682 103823-488 77270-864 77270-866 77310-286
2-Chloro-5-pyrimidineboronic acid 98%
2-Chloro-5-pyrimidineboronic acid

Precaución: for Research Use Only

Formula: C₄H₄BClN₂O₂
MW: 158.35 g/mol
Boiling Pt: 425.3 °C (760 mm Hg)
Flash Pt: 211.051 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08063113
CAS Number: 1003845-06-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR