2-Chloro-5-nitrophenylboronic acid 97%

Supplier: Ambeed
Warning

A129669-10G A129669-1G A129669-25G A129669-5G
103801-308EA 10 USD
103801-308 77109-148 77313-690 77313-692 77313-694
2-Chloro-5-nitrophenylboronic acid 97%
2-Chloro-5-nitrophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₅BClNO₄
Boiling Pt: 392.9 °C (760 mm Hg)
Melting Pt: 236…238 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03095323
CAS Number: 867333-29-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR