2-Chloro-5-nitrobenzo[d]thiazole 97%
Supplier: Ambeed
|
Warning
|
A166200-250MG
A166200-1G
A166200-5G
103806-022EA
84
USD
103806-022
103844-282
76784-060
77300-808
77300-810
2-Chloro-5-nitrobenzo[d]thiazole 97%
2-Chloro-5-nitrobenzo[d]thiazole
Precaución: for Research Use Only
Formula:
C₇H₃ClN₂O₂S Boiling Pt: 348.1 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD09540490 CAS Number: 3622-38-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...