2-Chloro-5-nitrobenzene-1-sulfonyl chloride 95%

Supplier: Ambeed
Danger

A456388-1G A456388-25G A456388-5G A456388-100MG A456388-10G A456388-250MG
103827-858EA 8 USD
103827-858 103838-934 76842-784 76842-786 76842-788 76979-516 76979-518 76979-520
2-Chloro-5-nitrobenzene-1-sulfonyl chloride 95%
2-Chloro-5-nitrobenzene-1-sulfonyl chloride

Precaución: For research use only

Formula: C₆H₃Cl₂NO₄S
MW: 256.06 g/mol
Boiling Pt: 371.6 °C (760 mm Hg)
Melting Pt: 88…90 °C
Flash Pt: 178.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00129813
CAS Number: 4533-95-3
UN: 3261
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR