4-Amino-2-chloro-5-methylpyrimidine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-5-methylpyrimidin-4-amine

A129810-10G A129810-250MG A129810-1G
103801-332EA 25 USD
103801-332 77285-268 77285-270 77313-722
4-Amino-2-chloro-5-methylpyrimidine 98%
4-Amino-2-chloro-5-methylpyrimidine

Precaución: for Research Use Only

Formula: C₅H₆ClN₃
MW: 143.58 g/mol
Boiling Pt: 334.8 °C (760 mm Hg)
Melting Pt: 232…235 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02683249
CAS Number: 14394-70-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR