2-Chloro-5-methylpyridine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-5-picoline

A150933-100G A150933-500G A150933-25G A150933-5G
103842-360EA 7 USD
103842-360 77110-294 77110-296 77299-840 77317-894
2-Chloro-5-methylpyridine 98%
2-Chloro-5-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₆ClN
MW: 127.57 g/mol
Boiling Pt: 190.8 °C (760 mm Hg)
Flash Pt: 90.55 °C
Storage Temperature: Ambient
MDL Number: MFCD00792460
CAS Number: 18368-64-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR