3-Amino-2-chloro-5-methylpyridine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-5-methylpyridin-3-amine

A407984-1G A407984-25G A407984-5G A407984-100G A407984-10G
103827-174EA 5 USD
103827-174 103838-218 76841-720 76841-722 76841-724 77115-784 77115-786
3-Amino-2-chloro-5-methylpyridine 98%
3-Amino-2-chloro-5-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₇ClN₂
MW: 142.5900 g/mol
Boiling Pt: 281.5 °C (760 mm Hg)
Melting Pt: 82…87 °C
Flash Pt: 124.1 °C
Storage Temperature: Ambient
MDL Number: MFCD03427656
CAS Number: 34552-13-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR