2-Chloro-5-methylpyrazine 97%
Supplier: Ambeed
|
Warning
|
A102645-100MG
A102645-10G
A102645-250MG
A102645-5G
103796-398EA
50
USD
103796-398
103822-096
77257-288
77257-290
77257-292
77308-970
2-Chloro-5-methylpyrazine 97%
2-Chloro-5-methylpyrazine
Precaución: for Research Use Only
Formula:
C₅H₅ClN₂ MW: 128.5600 g/mol Boiling Pt: 172 °C (760 mm Hg) Flash Pt: 72 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00055039 CAS Number: 59303-10-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...