6-Chloro-m-toluic acid 97%
Supplier: Ambeed
|
Warning
|
A374649-1G
A374649-25G
A374649-5G
A374649-100G
A374649-10G
103826-556EA
5
USD
103826-556
103837-574
76840-854
76840-856
76840-858
77115-424
77115-426
6-Chloro-m-toluic acid 97%
6-Chloro-m-toluic acid
Precaución: for Research Use Only
Formula:
C₈H₇ClO₂ Boiling Pt: 291.3 °C (760 mm Hg) Melting Pt: 148…151 °C Flash Pt: 130 °C Storage Temperature: Ambient |
MDL Number:
MFCD00045798 CAS Number: 6342-60-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...